Learn More
Hexane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.9% (>60% n-Hexane), Solstice
Shop All Honeywell Research Chemicals ProductsSpecifications
Specifications
| CAS Min % | >60.0000% |
| CAS Max % | >60.00%, <40.0000% |
| Density | 0.659 - 0.673 g/cm3 |
| Quantity | 19 L |
| Flash Point | −26°C |
| Packaging | Metal Can |
| UN Number | UN1208 |
| Water | 0.01% max. |
| Beilstein | 1730733 |
| Boiling Point | 68.7°C |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.