Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexane, Analytical Standard, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018167 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8167 50 mL
11-101-9017 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8167 Supplier MilliporeSigma Supelco Supplier No. 5275050ML
Add to Cart
Add to Cart

Specifications

CAS 110-54-3
Vapor Pressure 5.2 psi (37.7°C)
Vapor Density ˜3 (vs. air)
Density 0.659 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3(CH2)4 CH3
MDL Number MFCD00009520
Melting Point -95°C
Boiling Point 69°C
Synonym n-Hexane
Quantity 50 mL
Formula Weight 86.18
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.