Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexane CHROMASOLV™, for HPLC, ≥97.0% (GC), Solstice
GSA_VA

Catalog No. 6001046 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-010-46 1 L
60-010-52 6 x 1 L
60-010-50 4 x 2.5 L
60-010-51 4 x 4 L
60-010-47 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-010-46 Supplier Solstice Supplier No. 348591L
Add to Cart
Add to Cart

CHROMASOLV™, for HPLC,≥97.0% (GC)

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Density 0.659g/mL at 25°C
Quantity 1 L
Refractive Index n20/D 1.375
Linear Formula CH3(CH2)4CH3
Vapor Pressure 256mmHg (37.7°C)
Beilstein 1730733
Boiling Point 69°C
Formula Weight 86.18g/mol
Percent Purity ≥97.0% (GC)
Grade CHROMASOLV
Chemical Name or Material Hexane, for HPLC
Show More Show Less
Hazard Statement P201-P210-P273-P301 + P310-P308 + P313-P331
Precautionary Statement H225-H304-H315-H336-H361f-H373-H411
EINECSNumber 203-777-6
RTECSNumber MN9275000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.