Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, 99.9%, Extra Dry, mixture of isomers, AcroSeal™

Catalog No. AC610370010
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC610370010 1 L AcroSeal™ Glass Bottle
AC610371000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC610370010 Supplier Thermo Scientific Chemicals Supplier No. 610370010
Add to Cart

CAS: 92112-69-1 | C6H14

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexanes
CAS 92112-69-1, 110-54-3
Density 0.6590g/mL
Quantity 1 L
Identification Pass Test
Packaging AcroSeal™ Glass Bottle
Assay Percent Range ≤97%
Molecular Formula C6H14
Vapor Pressure Vapor Pressure: 160mbar at 20°C
Linear Formula CH3(CH2)4CH3
MDL Number MFCD00009520
Water 50ppm max.
Beilstein 01, 142
Merck Index 14, 4694
Synonym Hex
Formula Weight 86.18
Percent Purity ≤97%
Grade Extra Dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.