Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, technical, mixture of isomers

Catalog No. ac444340050
Change view
Click to view available options
Quantity:
5 L
Packaging:
HDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC444340050 5 L HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC444340050 Supplier Thermo Scientific Chemicals Supplier No. 444340050
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 92112-69-1 | C6H14 | 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexanes
CAS 92112-69-1, 110-54-3
Melting Point -95.0°C
Density 0.6590g/mL
Boiling Point 69.0°C
Flash Point −22°C
Packaging HDPE Bottle
Molecular Formula C6H14
Refractive Index 1.3748 to 1.3810
Linear Formula CH3(CH2)4CH3
Quantity 5 L
Specific Gravity 0.659
Synonym Hex
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Molecular Weight (g/mol) 86.18
Viscosity 0.31 mPa.s (20°C)
Formula Weight 86.18
Grade Technical
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.