Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, ACS Reagent, Reagents

Catalog No. 01337937
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Glass Bottle
Poly Bottle
Safety Coated Glass
Steel Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-337-937 4 L Glass Bottle
01-337-939 20 L Steel Pail
01-337-938 4 L Poly Bottle
01-337-936 4 L Safety Coated Glass
01-337-935 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-337-937 Supplier Reagents Holdings Llc Supplier No. C12139004C
Add to Cart
Add to Cart

Hexanes, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.665
Boiling Point 68.7°C
Grade ACS
Chemical Name or Material Hexanes, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.