Learn More
Specifications
Specifications
| Color | Colorless |
| Quantity | 4 L |
| Flash Point | -6.7°C (+20°F) |
| Identification | Pass Test |
| Packaging | Safe-Cote™ Amber Glass |
| Physical Form | Liquid |
| Residue after Evaporation | 0.001% max. |
| Specific Gravity | Less-than water |
| Assay Percent Range | ≥98.5 % |
| UN Number | 1208 |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. NFPA Health:1 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.