Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, HPLC grade, Ricca Chemical
SDP

Catalog No. RSOH001125D Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RSOH001125D 2.5 L
RSOH00111C 1 L
RSOH00114C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RSOH001125D Supplier Ricca Chemical Company Supplier No. RSOH00112.5D1
Add to Cart
Add to Cart

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name hexane
SMILES CCCCCC

Specifications

Absorbance 0.00 to 1.00, max. 0.25, max. 0.08, max. 0.01
CAS Max % 100
Appearance Colorless
Quantity 2.5 L
Packaging Glass Bottle
Residue after Evaporation max. 0.0005%
Titratable Acid max. 0.0003%
Water max. 0.02%
Boiling Point 69°C
Percent Purity min. 98.5%
Grade HPLC
Chemical Name or Material Hexanes
Show More Show Less
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.