Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HexArmor™ PointGuard™ Ultra 6044 Gloves

Catalog No. 19161136E Shop All Performance Fabrics, Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-161-136E X-Large
19-161-136A X-Small
19-161-136B Small
19-161-136C Medium
19-161-136D Large
19-161-136F 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-161-136E Supplier HexArmor™ Supplier No. 6044XL 10
Add to Cart
Add to Cart
This item is not returnable. View return policy

SuperFabric™ brand material offers highest level of cut-resistance on enhanced areas. Recommended for use as an under-glove solution with appropriate top-glove combination

  • High needlestick resistance provided by layering SuperFabric™ brand material
  • Tested using actual 25 gauge needles
  • Lightweight spandex shell with elastic wrist
  • Launderable

Specifications

Certifications/Compliance ANSI/ISEA 105 standard
Color Black
Cut Resistance Rating ANSI A9
Material SuperFabric
Product Type Cut-resistant Gloves
Size X-Large
EN388 Standard EN 388:4X33F
For Use With (Application) waste/recycle sorting, hospital waste handling, laundry and linen, wastewater/sewage treatment, municipality work
Glove Cuff Style Elasticated
Model 6044-XL
Series PointGuard™ Ultra 6044
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.