Description
Cut, puncture, and abrasion resistant aprons protect body from accidents on the job site. Help workers to stay warm, safe, and comfortable with less fatigue.
- Offer ISEA Level 5 cut resistance
- No metal disks or links to repair
- Lightweight—only 1/10 the weight of traditional chain mail safety wear
- Safe for machine wash and sanitizers
- Available in two lengths; both include adjustable strap
1These aprons are cut and/or puncture resistant, not cut or puncture proof. Always conduct a safe test with your application to determine if the product is suited for your needs.
Specifications
Specifications
| Size | Universal |
| Material | SuperFabric™ |
| Thickness | Single layer |
| Includes | Adjustable straps |
| For Use With (Application) | Meat packing, processing and general work |
| Certifications/Compliance | ISEA CE Level 5 |
| Dimensions (L x W) | 29 x 29 in. |