Learn More
Description
- IR-X™ Impact Exoskeleton™ with high-flex design keeps hands safe from smash & impact hazards
- Reinforced index finger and thumb crotch for increased longevity
- SlipFit™ cuff adds comfort and convenience
- Durable TP-X™ palm with reinforced stitching pattern
- Hi-vis color scheme for increased visual awareness on the job site
- Launderable
Specifications
Specifications
| Certifications/Compliance | ANSI/ISEA 138 Level 1 Impact Protection |
| Cuff Material | SlipFit™ |
| Color | Hi-vis Lime Green, Black |
| Cut Resistance Rating | ANSI A3 |
| Hand | Specific |
| Material | IR-X Impact Exoskeleton |
| Product Type | Mechanic's Gloves |
| Size | 2X-Large |
| For Use With (Application) | General Work, Extrication, Oil/Gas Operations, Machine Repair, and Mechanics |
| Puncture Rating | ASTM F2878 LEVEL 4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.