Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexatriacontane 98%, Thermo Scientific™

Catalog No. AC120730500
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC120730500 50 g
AC120730100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC120730500 Supplier Thermo Scientific Chemicals Supplier No. 120730500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-06-8
Molecular Formula C36H74
Molecular Weight (g/mol) 506.99
MDL Number MFCD00009413
InChI Key YDLYQMBWCWFRAI-UHFFFAOYSA-N
Synonym n-hexatriacontane, hexatriacontane, ch3-ch2 34-ch3, hexatriacontane, analytical standard, hexatriacontane, n, acmc-1b3b7, n-hexatriacontane 10g
PubChem CID 12412
ChEBI CHEBI:72688
IUPAC Name hexatriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 74°C to 78°C
Color White
Infrared Spectrum Authentic
Linear Formula CH3(CH2)34CH3
Quantity 50 g
Beilstein 01, I, 73
Formula Weight 506.97
Percent Purity ≥97.5%
Physical Form Shiny Flakes
Chemical Name or Material Hexatriacontane, 98%
EINECSNumber 211-127-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.