Learn More
Sigma Aldrich Fine Chemicals Biosciences Hexokinase from Saccharomy
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H63801.5KU
Encompass
Hexokinase (HK) from yeast is 486 amino acid containing enzyme with a α/β fold. It comprises a large and small domain separated by an active site cleft. Conserved glycine residues are essential for the ATP and glucose binding. The two isoforms are HKI and HKII encoded by HXK1 and HXK2 gene, respectively.
Catalog No. 50-289-1478
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.