Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl 2-methylbutanoate, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018972 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8972 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8972 Supplier MilliporeSigma Supelco Supplier No. 531841ML

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 10032-15-2
Density 0.857 g/mL (at 25°C)
Flash Point 84°C
Physical Form Neat
Linear Formula C2H5 CH(CH3)CO2(CH2)5 CH3
MDL Number MFCD00048880
Boiling Point 217°C to 219°C
Synonym Hexyl 2-methylbutyrate
Type Analytical Standard
Quantity 1 mL
Formula Weight 186.29
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.