Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl Acrylate (stabilized with HQ) 96.0+%, TCI America™
SDP

Catalog No. A125725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A125725ML 25 mL
A1257500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A125725ML Supplier TCI America Supplier No. A125725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2499-95-8
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00048881
InChI Key LNMQRPPRQDGUDR-UHFFFAOYSA-N
Synonym hexyl acrylate, n-hexyl acrylate, hexyl 2-propenoate, 2-propenoic acid, hexyl ester, n-hexylacrylate, ageflex n-ha, acrylic acid, hexyl ester, ccris 7038, acrylic acid-n-hexyl ester, hexylacrylate
PubChem CID 17259
IUPAC Name hexyl prop-2-enoate
SMILES CCCCCCOC(=O)C=C

Specifications

Color Yellow
Boiling Point 89°C
Quantity 25 mL
UN Number 3082
Formula Weight 156.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Acrylate (stabilized with HQ)
EINECSNumber (2)-3267
RTECSNumber AT1450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.