Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl chloroformate, 98%

Catalog No. p-3778754
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427040050 5 g Glass bottle
AC427040250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427040050 Supplier Thermo Scientific Chemicals Supplier No. 427040050
Only null left
Add to Cart

CAS: 6092-54-2 | C7H13ClO2 | 164.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6092-54-2
Molecular Formula C7H13ClO2
Molecular Weight (g/mol) 164.63
MDL Number MFCD00000651
InChI Key KIWBRXCOTCXSSZ-UHFFFAOYSA-N
Synonym hexyl chloroformate, carbonochloridic acid, hexyl ester, n-hexyl chloroformate, chloroformic acid n-hexyl ester, hexyl chlorocarbonate, unii-mu6004c2z3, chloro hexyloxy methanone, chloroformic acid hexyl ester, hexylchloroformate, hexyl=chloroformate
PubChem CID 22466
IUPAC Name hexyl carbonochloridate
SMILES CCCCCCOC(Cl)=O

Specifications

Color Colorless
Density 1.0070g/mL
Boiling Point 60°C to 61°C (7.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4230 to 1.4250 (20°C, 589nm)
Linear Formula ClCO2(CH2)5CH3
Quantity 5 g
Specific Gravity 1.007
Formula Weight 164.63
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Hexyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic if inhaled.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-036-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.