Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. T0883500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0883500G 500 g
T088325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0883500G Supplier TCI America Supplier No. T0883500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3839-35-8
Molecular Formula C13H20O3S
Molecular Weight (g/mol) 256.36
MDL Number MFCD00059439
InChI Key IVQOVYWBHRSGJI-UHFFFAOYSA-N
Synonym p-Toluenesulfonic Acid Hexyl Ester
PubChem CID 220448
IUPAC Name hexyl 4-methylbenzenesulfonate
SMILES CCCCCCOS(=O)(=O)C1=CC=C(C=C1)C

Specifications

Color Yellow
Boiling Point 197°C
Quantity 500 g
Formula Weight 256.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl p-Toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.