Learn More
HF Scientific™ Powder Pop™ Dispensers for Chlorine Testing
Catalog No. 15393150
Click to view available options
For Use With (Application):
For testing free chlorine
For testing total chlorine
For Use With (Equipment):
Hach and HF Scientific Photometers
Hach and Hanna Photometers
LaMotte, Orbeco-Hellige, YSI, Palintest, Tintometer, Orion Photometers
Sample Size:
5 mL
10 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) For Use With (Equipment) Sample Size
15-393-150 For testing free chlorine Hach and HF Scientific Photometers 5 mL
15-393-154 For testing free chlorine LaMotte, Orbeco-Hellige, YSI, Palintest, Tintometer, Orion Photometers 10 mL
15-393-155 For testing total chlorine LaMotte, Orbeco-Hellige, YSI, Palintest, Tintometer, Orion Photometers 10 mL
15-393-152 For testing free chlorine Hach and Hanna Photometers 10 mL
15-393-153 For testing total chlorine Hach and Hanna Photometers 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 15-393-150 Supplier HF Scientific™ Supplier No. 10500
Add to Cart
Add to Cart
For testing free or total chlorine
  • Easy to dispense into round or square cuvettes
  • Each supplied with enough powder for 100 doses
  • Available for 5, 10 or 25mL sample volumes
  • Simple push button operation
Specifications
For Use With (Application) For testing free chlorine
For Use With (Equipment) Hach and HF Scientific Photometers
No. of Tests 100
Sample Size 5 mL
Type Dispenser