Learn More
Erlab™ HF2.BE Carbon Filter For Captair Storage Cabinets 822 and 832
Carbon filter for use with storage cabinets 822 and 832 for the removal of acids, organics, inorganics and bases.
Supplier: Erlab™ HF2.BE
Safety Services Available
Overview
Risk Assessment
This item is not returnable.
View return policy
Description
Replacement filters for CaptairStore Filtered Storage Cabinets 822 and 832.
Specifications
Carbon | |
4 in. | |
5.9 Kg | |
2 in. | |
380 mm | |
Carbon Filter | |
Acid and Organic Vapor Filtering |
CaptairStore Cabinets 822 and 832 | |
13 lb. | |
15 in. | |
102 mm | |
482 mm | |
0/1 V |