Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erlab™ HF2.HP HEPA Filter For Captair Storage Cabinets 822 and 832
GREENER_CHOICE

HEPA H14 filter providing 99.995% efficiency for trapping particles 0.1 micron or higher.

Supplier:  Erlab™ HF2.HP

Catalog No. 08-444-078


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

Replacement filters for CaptairStore Filtered Storage Cabinets 822 and 832.

Specifications

Specifications

HEPA Filter
HEPA
Polypropylene
Black
2 in.
HF2.HP Filter
0/1 V
5.4 kg
406 mm
Powder
LED
Protector Airo
CaptairStore Cabinets 822 and 832
50 mm
None
12 lb.
16 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.