Learn More
MyBioSource Human Secondary Antibody
SDP
Supplier:  MyBioSource MBS85748171 MG
Encompass_Preferred
The product is a human secondary antibody from MyBioSource. It is polyclonal and was produced in rabbit using purified human immunoglobulin as the immunogen. The antibody can detect human IgG by ELISA, WB, IHC, and FC applications. It is supplied as a lyophilized powder with BSA and sodium azide as stabilizers. The antibody should be stored at -20C. The secondary antibody allows detection of primary antibodies specific for human antigens. It can be used in research applications to study human proteins. The secondary antibody binds the Fc region of human IgG.
Catalog No. NC2669990