Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HFE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588983
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588983 Supplier Invitrogen™ Supplier No. PA588983
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSQMWLQLSQ SLKGWDHMFT VDFWTIMENH NHSKESHTLQ VILGCEMQED NSTEGYWKYG YDGQDHLEFC PDTLDWRAAE PRAWPTKLEW ERHKIRARQN RAYLERDCPA QLQQLLELGR GVLDQQVPPL VKVTHHVTSS VTTLRCRALN YYPQNITMKW LKDKQPMDAK EFEPKDVLPN GDGTYQGWIT LAVPPGEEQR YTCQVEHPGL DQPLIVIWEP SPSGTLV; Positive Samples: Mouse liver, Mouse heart; Cellular Location: Cell membrane, Single-pass type I membrane protein.

The protein encoded by this gene is a membrane protein that is similar to MHC class I-type proteins and associates with beta2-microglobulin. It is thought that this protein functions to regulate iron absorption by regulating the interaction of the transferrin receptor with transferrin. The iron storage disorder, hereditary haemochromatosis, is a recessive genetic disorder that results from defects in this gene. At least nine alternatively spliced variants have been described for this gene. Additional variants have been found but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HFE
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.40 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HFE
Gene Accession No. P70387, Q30201
Gene Alias hemochromatosis; hereditary hemochromatosis protein; hereditary hemochromatosis protein HLA-H; hereditary hemochromatosis protein homolog; Hfe; HFE 1; HFE1; HH; high Fe; HLA H; HLAH; HLA-H; MGC:150812; MGC10379; MGC103790; MHC class I-like protein HFE; Mr2; MVCD7; RT1-CAFE; TFQTL2
Gene Symbols HFE
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 80-306 of human HFE (NP_0004011).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15216, 3077
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.