Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ HG-9-91-01

Catalog No. 898550
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-855-0 50 mg
89-851-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-855-0 Supplier Tocris Bioscience™ Supplier No. 8985/50
Only null left
Add to Cart
Add to Cart

Potent pan-salt inducible kinases (SIK1, SIK2 and SIK3) inhibitor. Also inhibits Lat-B-induced EP differentiation via nuclear YAP1 accumulation.

HG-9-91-01 (SIK1 inhibitor) is a potent pan-salt inducible kinases (pan-SIKs) inhibitor (IC50 values are 0.92, 6.6 and 9.6 nM for SIK1, SIK2 and SIK3 respectively). HG-9-91-01 inhibits latrunculin B (Lat-B)-induced endocrine progenitors (EP) differentiation via nuclear YAP1 accumulation in vitro. HG-9-91-01 increases LPS-stimulated IL-10 production and suppresses pro-inflammatory cytokine secretion in vitro. HG-9-91-01 increases β-cell proliferation in vitro and in vivo (mouse, zebrafish).

Specifications

Chemical Name or Material 1-(2,4-Dimethoxyphenyl)-3-(2,6-dimethylphenyl)-1-(6-((4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)urea
CAS 1456858-58-4
Quantity 50 mg
Target Other Kinase Inhibitors
Molecular Formula C32H37N7O3
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.