Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HGF Monoclonal Antibody (SBF5)
GREENER_CHOICE

Catalog No. PIMA514160 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514160 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514160 Supplier Invitrogen™ Supplier No. MA514160
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA5-14160 targets Hepatocyte Growth Factor in WB and ICC/IF applications and shows reactivity with Human and Mouse samples. The MA5-14160 immunogen is recombinant human HGF protein.

Hepatocyte growth factor regulates cell growth, cell motility, and morphogenesis by activating a tyrosine kinase signaling cascade after binding to the proto-oncogenic c-Met receptor. Hepatocyte growth factor is secreted by mesenchymal cells and acts as a multi-functional cytokine on cells of mainly epithelial origin. Its ability to stimulate mitogenesis, cell motility, and matrix invasion gives it a central role in angiogenesis, tumorogenesis, and tissue regeneration. It is secreted as a single inactive polypeptide and is cleaved by serine proteases into a 69-kDa alpha-chain and 34-kDa beta-chain. A disulfide bond between the alpha and beta chains produces the active, heterodimeric molecule. The protein belongs to the plasminogen subfamily of S1 peptidases but has no detectable protease activity. Alternative splicing of this gene produces multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HGF
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone SBF5
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Hgf
Gene Accession No. P14210, Q08048
Gene Alias C230052L06Rik; DFNB39; Fibroblast derived tumor cytotoxic factor; fibroblast-derived tumor cytotoxic factor; F-TCF; hepatocyte growth factor; Hepatocyte growth factor (hepapoietin A; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoeitin A; hepatopoeitin-A; HepatopoietinA; hepatopoietin-A; Hgf; HGF/SF; HGFB; HPTA; Lung fibroblast derived mitogen; lung fibroblast-derived mitogen; NK1; NK2; Scatter factor; scatter factor); SF; SF/HGF
Gene Symbols Hgf
Host Species Mouse
Immunogen Recombinant human HGF protein.
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15234, 3082
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.