Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HGK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104232
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5104232 100 μL
1 options

Catalog No. PIPA5104232

Supplier: Invitrogen™ PA5104232

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MAP4K4.

MAP4K4 (HGK) is a serine/threonine kinase which plays an important role in stress-activated MAPK core signaling pathways, the response to environmental stress and cytokines such as TNF-alpha. It appears to act upstream of the JUN N-terminal pathway. This protein is thought to interact with the SH3 domain of the adapter proteins Nck. HGK binds, via its CNH regulatory domain, to the N-terminal region of SPG3A. Expression appears to be ubiquitous, expressed in all tissue types examined. Isoform 5 appears to be more abundant in the brain, and isoform 4 is predominant in the liver, skeletal muscle and placenta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HGK
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAP4K4
Gene Accession No. O95819, P97820
Gene Alias 9430080K19Rik; AU043147; AU045934; AW046177; epididymis secretory protein Li 31; FLH21957; FLJ10410; FLJ20373; FLJ90111; HEL-S-31; hepatocyte progenitor kinase-like/germinal center kinase-like kinase; HGK; HPK/GCK-like kinase HGK; KIAA0687; Map4k4; MAPK/ERK kinase kinase kinase 4; MEK kinase kinase 4; MEKKK 4; MEKKK4; mitogen activated protein kinase kinase kinase kinase 4; mitogen-activated protein kinase kinase kinase kinase 4; NCK interacting kinase; nck-interacting kinase; NIK; Ste20 group protein kinase HGK
Gene Symbols MAP4K4
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAP4K4(Accession O95819), corresponding to amino acid residues E410-R460.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26921, 301363, 9448
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.