Learn More
Medchemexpress LLC Benzo[b]thiophene-2-carboxamide, 3-chloro-4,7-difluoro-N-[trans-4-(methylamino)cyclohexyl]... | 612542-14-0 | 99.9% | 526.04 | C28H26ClF2N3OS | 100MG
SDP
Supplier:  Medchemexpress LLC HY124899100MG
Encompass_Preferred
Hh-Ag1.5 is a potent small-molecule agonist of the Hedgehog signaling pathway that targets Smoothened with an EC50 of about 1 nM. Supplied as a high-purity solid, it is intended for research use in both in vitro and in vivo studies of Hedgehog signaling. The compound exhibits high solubility in DMSO and has defined storage conditions for long-term stability.
  • Potent Hedgehog pathway agonist with EC50 ≈ 1 nM.
  • High purity suitable for analytical and biological assays.
  • Soluble in DMSO at approximately 50 mg/mL for in vitro work.
  • Formulatable for in vivo dosing in common vehicle mixtures.
  • Stable as a powder under frozen storage for extended periods.
  • Available in multiple pack sizes to support screening and scale-up.
Catalog No. 50-002-37035