Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-chloro-4,7-difluoro-N-[4-(methylamino)cyclohexyl]-N-[(3-pyridin-4-ylphenyl)methyl]-1-benzothiophen | 612542-14-0 | 99.9% | 526.04 | C28H26ClF2N3OS | 50MG
SDP

Supplier:  Medchemexpress LLC HY12489950MG

Encompass_Preferred

Hh-Ag1.5 is a potent small-molecule agonist of the Hedgehog signaling pathway (EC50 ≈ 1 nM) for use in in vitro and in vivo pathway activation and stem-cell reprogramming studies. Supplied as a white to off-white solid at high purity, it is formulated for research and preclinical applications.

  • Potency: EC50 1 nM (Hedgehog)
  • CAS number: 612542-14-0
  • Molecular formula: C28H26ClF2N3OS
  • Molecular weight: 526.04
  • Purity: 99.9%
  • Appearance: white to off-white solid
  • Solubility: DMSO 50 mg/mL (use fresh DMSO; ultrasonic may be required)
  • Storage: powder -20°C (3 years), 4°C (2 years)
  • Typical pack sizes: 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg

Catalog No. 50-002-35947


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.