Learn More
ATCC HH; T Cell Lymphoma; Human
Supplier:  ATCC CRL2105
Encompass
HH are T lymphoblasts that were isolated from the peripheral blood of a 61-year-old White male with cutaneous T cell lymphoma. It has applications in immuno-oncology and immunology.
Catalog No. 50-238-2550