Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HHATL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534355
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534355 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534355 Supplier Thermo Scientific Supplier No. PA534355
Only null left
Add to Cart
Add to Cart

HHATL Polyclonal Antibody

HHATL Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen HHATL
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene HHATL
Gene Accession No. Q9D1G3, Q9HCP6
Gene Alias 1110011D13Rik; C3orf3; Glycerol uptake/transporter homolog; GUP1; GUP1 glycerol uptake/transporter homolog; Gup1, glycerol uptake/transporter homolog; hedgehog acyltransferase-like; hedgehog acyltransferase-like protein; HHATL; KIAA1173; MBOAT3; membrane bound O-acyltransferase domain containing 3; MSTP002; OACT3; Protein-cysteine N-palmitoyltransferase HHAT-like protein; RGD1311911
Gene Symbols HHATL
Host Species Rabbit
Immunogen A 19 amino acid synthetic peptide near the amino terminus of human HHATL.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301073, 57467, 74770
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.