Learn More
Jackson Safety HHIS Face Shield Hard Hat Adaptor for Quad™ 500 Series 14225 and 14235
The Jackson Safety™ Hard Hat Interchange System lets you add, remove, or switch between a face shield or welding helmet all without having to remove your head protection.
Supplier:  Jackson Safety 38423
Catalog No. 19-060-795
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Hard Hat Interchange System for Slotted Hard Hats
  • Replacement HHIS Knobs for 14425/14235/14238 QUAD 500
Specifications
Specifications
Slotted Har Hats, Welding Helmets, Faceshields
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.