Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syngene™ Hi Intensity LEDS for GBOX
GREENER_CHOICESDP

Catalog No. 01257282 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Color:
Green, Blue, Far Red
Green, Blue, Infa Red, Far Red
Infa Red, Far Red
Red, Green, Blue
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01-257-282 Green, Blue, Infa Red, Far Red
01-257-280 Green, Blue, Far Red
01-257-278 Infa Red, Far Red
01-257-276 Red, Green, Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-257-282 Supplier Syngene™ Supplier No. HILEDGBIRFR
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Syngene™ Hi Intensity LEDS for GBOX includes gantry, electronics and lighting units, and works with latest GBOX ONLY (not previous modular system).

  • For use with GBOX
  • Includes gantry, electronics and lighting units

Specifications

For Use With (Application) Multiplex Western Fluorescence
Quantity 1
Color Green, Blue, Infa Red, Far Red
For Use With (Equipment) Gbox Chemi Systems
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.