Learn More
Specifications
Specifications
| For Use With (Application) | Multiplex Western Fluorescence |
| Quantity | 1 |
| Color | Red, Infra Red |
| For Use With (Equipment) | Gbox Chemi Systems |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our missing translation for 'privacyPolicy'.

