Learn More
PIP Hi-Vis Apparel Vest Yellow
Catalog No. 17033744 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-033-744 Medium
17-033-745 Large
17-033-746 X-Large
17-033-747 2X-Large
17-033-748 3X-Large
17-033-749 4X-Large
17-033-750 5X-Large
17-033-743 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-033-744 Supplier PIP Supplier No. 3020500YEL/M
Only null left
Add to Cart
Add to Cart
ANSI Type R Class 2 Two-Tone Eleven Pocket Surveyors Vest with Solid Front and Mesh Back
  • Solid front and mesh back for breathability
  • 2 inch two-tone tape for optimum visibility
  • Zipper closure
  • 2 mic tabs with reinforced stitching
  • 2 lower external flap pockets
  • 2 internal lower pockets
  • 1 chest radio pocket
  • 1 chest pocket with 4 pen pockets in front
Specifications
Product Type High-visibility Vest
Construction Solid Front, Mesh Back
Closure Type Zipper
Color Hi-vis Yellow
Size Medium
Material Polyester
No. of Pockets 11