Learn More
Wells Lamont™ Hi-Vis Nitrile-Coated Impact Resistant Work Gloves with Fleece Lining
Shop All Wells Lamont Industry Group ProductsDescription
- Impact resistant work gloves deliver an extra level of protection to the back of the hand and fingers
- Hi-vis yellow shell constructed of a synthetic fiber blend improves awareness on the job site
- Sandy nitrile coating on the palm provides a non-slip surface in wet, dry and oily conditions
- Fleece liner increases warmth in cold conditions
- Ideal for applications that require multi-hazard protection, grip, dexterity, and thermal protection
Specifications
Specifications
| Coating Material | Nitrile |
| Color | Hi-vis Orange |
| Cut Resistance Rating | ANSI A4 |
| Hand | Specific |
| Material | HPPE |
| Size | Medium |
| Glove Cuff Color | Hi-Vis Yellow |
| Glove Cuff Style | Knit |
| Liner Material | Fleece |
| Series | I2449T |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.