Learn More
MCR Safety™ Hi-Vis Reflective Lime Mesh Safety Vest, 5 pt. break, CSA Cl. 2
Catalog No. 19034363 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-034-363 Small
19-034-365 Medium
19-034-366 Large
19-034-358 X-Large
19-034-362 2X-Large
19-034-361 3X-Large
19-034-360 4X-Large
19-034-359 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-034-363 Supplier MCR Safety™ Supplier No. RXCL2MLS
Only null left
Add to Cart
Add to Cart
Luminator™ Series Hi-Vis reflective lime safety vest - manufactured with recycled materials, meets ANSI/ISEA 107 2020 standard, type R class 2 compliant, five point break away design.
  • Meets ANSI/ISEA 107 2020 Standard - Type R, Class 2 compliant
  • 5 Point break away pattern
  • CSA Z96 Class 2 Level 2 compliant
  • D-Ring access slot on back
  • Hook and Loop front closure
  • Fluorescent lime mesh polyester material
  • 1 outer chest pocket
  • Made of recycled materials
  • 2 Inch silver reflective stripes
  • X reflective pattern on vest back
Specifications
Closure Type Breakaway Hook and Loop
Product Type High-visibility Vest
Color Hi-vis Lime
Size Small
Material Polyester Mesh
No. of Pockets 1