Learn More
Pyramex Safety Products Hi-Vis Safety Vest with Contrasting Reflective Tape
Catalog No. 19555239 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Lime
Orange
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-555-239 Lime Large
19-555-240 Lime Medium
19-555-245 Lime X-Large
19-555-241 Lime 2X-Large
19-555-242 Lime 3X-Large
19-555-244 Lime 5X-Large
19-555-247 Orange Medium
19-555-246 Orange Large
19-555-252 Orange X-Large
19-555-248 Orange 2X-Large
19-555-249 Orange 3X-Large
19-555-250 Orange 4X-Large
19-555-251 Orange 5X-Large
19-555-243 Lime 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 19-555-239 Supplier Pyramex Safety Products Supplier No. RCZ2410L
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex Safety™ Hi-Vis Safety Vest with Contrasting Reflective Tape features a 4 inch. contrasting color trim around bottom and over shoulders which form an X on the back.
Specifications
Closure Type Zipper
Color Lime
Size Large
Material Solid Polyester Front, Mesh Back
Includes 2 mic holder loops
No. of Pockets 8: 5 outside, 3 inside
Tape Type 2 in. silver reflective tape, 4 in. contrasting color trim