Learn More
Molnlycke Hibiclens™ Skin Cleanser
Description
- With multiple indications, Hibiclens has a great deal of utility throughout the hospital facility. The 4% chlorhexidine gluconate solution is in a gentle, skin cleansing base that can be used for:
- Daily patient bed baths, as part of a horizontal infection prevention approach
- Preoperative/postoperative showering and decolonization, as part of surgical site infection prevention bundles
- Surgical hand scrubbing
- Personnel hand washing
- Preoperative skin preparation
Specifications
Specifications
| Product Type | Antiseptic Skin Cleanser |
| Form | Liquid |
| For Use With (Application) | Daily Patient Bed Baths, Preoperative/Postoperative Showering, Surgical Hand Scrubbing, Personnel Hand Washing, Preoperative Skin Preparation |
| Quantity | 12/Cs. |
| Volume (English) | 32 oz. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.