Learn More
Applied Biosystems™ HID Real-Time PCR Analysis Software v1.2
Description
HID Real-Time PCR Analysis Software is a tool for forensic laboratories to optimize their quantification workflow and enhance their downstream decision making process.
The software has the following workflow features:
- customizable sample templates
- advanced data analysis capabilities
- normalization tool
- STR setup tool
HID Real-Time PCR Analysis Software v1.2 is designed for the Applied Biosystems™ 7500 Real-Time PCR System. It offers a set of features beyond instrument controls. It streamlines the experimental setup, supports customizable workflow templates and enhances the data analysis and review process. The software is validated for use on both the Windows™ XP and Windows™ 7 platforms, and it is specialized for all available Quantifiler™ kits, including the Quantifiler™ Trio and the Quantifiler™ HP DNA Quantification kits.
This product includes an HID Real-Time PCR Analysis Software v1.2 DVD and 5 user licenses. Documentation is not included.
Specifications
Specifications
| For Use With (Equipment) | 7500 System |
| Software Type | 1 User License (New Install) |
| Product Type | HID Real Time PCR Analysis Software |
| Quantity | 1 License |
For Research Use Only. Not for use in diagnostic procedures.