Learn More
Applied Biosystems™ HID Real-Time PCR Analysis Software v1.3
Description
HID Real-Time PCR Analysis Software is a tool that helps enable forensic laboratories to optimize their quantification workflow and enhance their downstream decision-making process.
The software has the following workflow features:
- Virtual standard curve functionality
- Customizable templates
- Advanced data analysis capabilities including quality flags
- STR sample normalization (dilution) and reaction setup tool
HID Real-Time PCR Analysis Software v1.3 is designed for use with the Applied Biosystems 7500 and QuantStudio 5 real-time PCR systems. It offers a set of features beyond instrument controls. HID Real-Time PCR Analysis Software v1.3 streamlines the experimental setup, supports customizable workflow templates, and enhances the data analysis and review process. It is validated for use on the Windows™ 7 platform and is may be used with all available HID Quantifiler kits
This product includes an HID Real-Time PCR Analysis Software v1.3 DVD and 1 user license. Documentation is not included.
Specifications
Specifications
| Software Type | Real-Time PCR Analysis |
| Quantity | 1 License |
| For Use With (Equipment) | Applied Biosystems 7500, QuantStudio 5 Real-Time PCR System |
| Product Type | HID Real-Time PCR Analysis Software V1.3 |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.