Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HIF-1 alpha Antibody Pack
SDP

Catalog No. NB100905 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100905 5 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100905 Supplier Novus Biologicals™ Supplier No. NB100905
Add to Cart
Add to Cart

Includes

  • 1.NB100-105: HIF-1 alpha Antibody (H1alpha67)
  • 2.NB100-131: HIF-1 alpha Antibody (ESEE122)
  • 3.NB100-449: HIF-1 alpha Antibody
  • 4.NB100-479: HIF-1 alpha Antibody
  • 5.NB100-654: HIF-1 alpha Antibody

For Research applications

HIF-1 alpha specifically detects HIF-1 alpha in Canine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot.

Specifications

Antigen HIF-1 alpha
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot
Conjugate Unconjugated
Dilution Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp
Gene Symbols HIF1A
Immunogen For NB 100-654: Human HIF-1 alpha amino acids 432-528., For NB 100-105: Human HIF-1 alpha amino acids 432-528., For NB 100-131: Human HIF-1 alpha amino acids 329-530., For NB 100-449: Human HIF-1 alpha amino acids 775-826., For NB 100-479: Antibody was developed against a fusion protein including amino acids 530 - 825 of the mouse HIF-1 alpha protein [Uniprot# Q61221].
Quantity 5 Vials
Research Discipline Cancer, HIF Target Genes, Hypoxia, mTOR Pathway
Gene ID (Entrez) 3091
Test Specificity These antibodies are specific for HIF-1alpha.
Includes 1.NB100-105: HIF-1 alpha Antibody (H1alpha67)
2.NB100-131: HIF-1 alpha Antibody (ESEE122)
3.NB100-449: HIF-1 alpha Antibody
4.NB100-479: HIF-1 alpha Antibody
5.NB100-654: HIF-1 alpha Antibody
Target Species Canine, Human, Rat, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.