Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HIF-1 alpha Antibody Packs
SDP

Catalog No. NB100901IHC Shop All Novus Products
Change view
Click to view available options
Quantity:
4 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100901IHC 4 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100901IHC Supplier Novus Biologicals™ Supplier No. NB100901IHC

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

For Research applications

Specifications

Antigen HIF-1 alpha
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown
Conjugate Unconjugated
Dilution Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated
Gene Alias AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp
Gene Symbols HIF1A
Immunogen NB100-131: Human HIF-1 alpha amino acids 329-530. NB100-123: Fusion protein containing amino acids 432-528 of human HIF-1alpha. NB100-449: peptide between residues 775 and the C-terminus (residue 826) of human HIF-1 alpha. NB100-479: A fusion protein including amino acids 530 - 825 of the mouse HIF-1 alpha protein [Uniprot# Q61221].
Quantity 4 Vials
Research Discipline Cancer, HIF Target Genes, Hypoxia, mTOR Pathway
Gene ID (Entrez) 3091
Test Specificity These antibodies are specific for HIF-1 alpha.
Includes 1.NB100-123: HIF-1 alpha Antibody (H1alpha67)
2.NB100-131: HIF-1 alpha Antibody (ESEE122)
3.NB100-449: HIF-1 alpha Antibody
4.NB100-479: HIF-1 alpha Antibody
Target Species Human, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.