Learn More
Invitrogen™ HIF-1 beta Recombinant Rabbit Monoclonal Antibody (3R0Y4)
Shop All Thermo Scientific Products

Description
Immunogen sequence: LAPRQQQQQT ELDMVPGRDG LASYNHSQVV QPVTTTGPEH SKPLEKSDGL FAQDRDPRFS EIYHNINADQ SKGISSSTVP ATQQLFSQGN TFPPTPRPAE NFRNSGLAPP VTIVQPSASA GQMLAQISRH SNPTQGATPT WTPTTRSGFS AQQVATQATA KTRTSQFGVG SFQTPSSFSS MSLPGAPTAS PGAAAYPSLT NRGSNFAPET GQTAGQFQTR TAEGVGVWPQ WQGQQPHHRS SSSEQHVQQP PAQQPGQPEV FQEMLSMLGD QSNSYNNEEF PDLTMFPPFS E.
Specifications
Specifications
| Antigen | HIF-1 beta |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 3R0Y4 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ARNT |
| Gene Accession No. | P27540 |
| Gene Alias | AHA-1; ARNT; ARNT protein; Arnt1; ARNT1a; ARNT1b; aryl hydrocarbon receptor nuclear translocater; Aryl hydrocarbon receptor nuclear translocator; Aryl hydrocarbon receptor nuclear translocator 1; aryl hydrocarbon receptor nuclear translocator type 1a; aryl hydrocarbon receptor nuclear translocator type 1b; bHLHe2; Class E basic helix-loop-helix protein 2; D3Ertd557e; dioxin receptor, nuclear translocator; Drnt; ESTM42; hif 1; HIF1 beta; HIF-1 beta; Hif1b; HIF1BETA; HIF-1beta; HIF-1-beta; HIF1-beta; hypoxia-inducible factor 1 beta; hypoxia-inducible factor 1, beta subunit; hypoxia-inducible factor 1-beta; mKIAA4051; TANGO; W08714; zgc:136664 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.