Learn More
Description
Includes
- 1.NB100-122: HIF-2 alpha/EPAS1 Antibody
- 2.NB100-132: HIF-2 alpha/EPAS1 Antibody (ep190b)
Specifications
Specifications
| Antigen | HIF-2 alpha/EPAS1 |
| Applications | ChIP Assay, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP), Knockdown Validated |
| Gene Alias | Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF-2 alpha, |
| Gene Symbols | EPAS1 |
| Host Species | Mouse |
| Immunogen | NB 100-122 = Proprietary peptide derived from the mouse/human HIF-2a. NB 100-132 = Human HIF-2a amino acids 535-631. |
| Quantity | 2 Vials |
| Research Discipline | Angiogenesis, Apoptosis, Cancer, Cancer Stem Cells, Chromatin Research, Embryonic Stem Cell Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
