Learn More
HIF1-alpha Recombinant Rabbit Monoclonal Antibody (BL-124-3F7), Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
Specifications
Specifications
| Antigen | HIF1-alpha |
| Applications | ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | BL-124-3F7 |
| Conjugate | Unconjugated |
| Formulation | BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 |
| Gene | HIF1A |
| Gene Accession No. | Q16665 |
| Gene Alias | Hypoxia inducing factor 1 |
| Gene Symbols | HIF1A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.