Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One High Altitude Coagulation Tubes with Sodium Citrate Solution
Click to view available options
Capacity (Metric):
1.8 mL
3.15 mL
Description
- For use in high altitude elevations above 5000 ft.
- 13 x 75 mm tubes filled with buffered tri-sodium citrate solution
- 3.2 % (0.109 mol/L) concentration
- Mixing ratio is one part citrate to nine parts blood when tube is filled appropriately as indicated by fill mark on label
- Clear as glass with equal or better performance
- Molded to be virtually unbreakable, providing a better level of safety than glass
- 18-month shelf life from date of manufacture
- Blue nonscrew safety pull cap can be removed with a gentle pull

Specifications
Specifications
Product Type | High Altitude Sodium Citrate Blood Collection Tube |
Additive | 3.2% Citrate Solution |
Material | PET |
Capacity (Metric) | 3.15 mL |
Closure Color | Blue with Black Ring |
Tube Size | 13 x 75 mm |
For Use With (Application) | High altitude elevations above 5000 ft. |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction