Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences High Fidelity DNA Polymerase 2X Master Mix
SCI_ED

Catalog No. S03148
Change view
Click to view available options
No. of Reactions:
100
500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
S03148 500
S03149 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03148 Supplier G-Biosciences Supplier No. 7861969
Add to Cart
Add to Cart

High Fidelity DNA Polymerase 2x Master Mix is used for PCR, Cloning, long and high GC content template DNA amplification and Diagnostics purposes.

  • G-biosciences high-fidelity DNA Polymerase 2x master mix is ready to use premix which contains high fidelity DNA Polymerase, dNTPs and optimized buffer with stabilizers.
  • The high-fidelity DNA Polymerase possesses 5' - 3' polymerase and 3' - 5' exonuclease (proofreading) activities which has the fidelity, sensitivity and processivity with an error rate ~2.8 x 102-fold lower than Taq DNA polymerase, and significantly lower than other proofreading DNA polymerases.

Specifications

Detection Method qPCR, RT-PCR
Form Liquid
GC-Rich PCR Performance High
PCR Method 1-step RT-qPCR, Multiplex qPCR, Standard PCR, End-point PCR, 1-step RT-PCR
Reaction Speed Fast
Product Type DNA Polymerase Master Mix
For Use With (Application) RT-PCR, Standard PCR, Cloning, Site-directed Mutagenesis, Multiplex PCR
No. of Reactions 500
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.