Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences High Fidelity DNA Polymerase
SCI_ED

Catalog No. S03151
Change view
Click to view available options
No. of Reactions:
100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
S03151 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03151 Supplier G-Biosciences Supplier No. 7861966
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High fidelity DNA Polymerase is used for PCR, cloning, long and high GC content template DNA amplification and diagnostics purposes.

  • G-biosciences high-fidelity DNA Polymerase possesses 5' - 3' polymerase and 3' - 5' exonuclease (proofreading) activities.
  • This Polymerase has the fidelity, sensitivity and processivity with an error rate ~2.8 x 102-fold lower than Taq DNA polymerase, and significantly lower than other proofreading DNA polymerases

Specifications

Detection Method Gel Electrophoresis
Form Liquid
GC-Rich PCR Performance High
PCR Method Standard PCR, Direct PCR, qPCR, Multiplex qPCR, 1-step RT-qPCR
Reaction Speed Fast
Label or Dye Biotin, 6-FAM, ROX, SYBR Green, TAMRA
Product Type DNA Polymerase
For Use With (Application) Sequencing, Multiplex PCR, High-fidelity PCR, Site-directed Mutagenesis, Next-Generation Sequencing (NGS)
No. of Reactions 100
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.