Learn More
High Five™ Disposable Face Masks
Catalog No. 19167934 Shop All Ansell Products
Change view
Click to view available options
Model:
AM101
Color:
Blue
Fastening Type:
Earloops
Catalog No. Model Color Fastening Type
19-167-934 AM101 Blue Earloops
1 options
This item is not returnable. View return policy
Catalog No. 19-167-934 Supplier High Five™ Supplier No. EB1059900
This item is not returnable. View return policy
Ideal for various medical or lab procedures.
  • Non-woven
  • 3-ply masks are designed for comfort
TRUSTED_SUSTAINABILITY
Specifications
Product Type Medical/Procedural Face Mask
Color Blue
Fastening Type Earloops
Material Polypropylene
Style Pleated
Disposable Single-use
Filter Efficiency 95% (BFE)
Latex Free Yes
Model AM101