Learn More
High Five™ Microporous Coveralls
Catalog No. 19167902 Shop All Ansell Products
Change view
Click to view available options
Description:
Elastic wrists and ankles
Elastic wrists; Attached hood and boots
Elastic wrists; Attached hood and boots;
Knit Collar and Cuffs; Attached hood and boots
Cuff Style:
Elastic
Knit
Size:
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Cuff Style Size
19-167-902 Elastic wrists and ankles Elastic Large
19-167-905 Elastic wrists and ankles Elastic 3X-Large
19-167-903 Elastic wrists and ankles Elastic X-Large
19-167-904 Elastic wrists and ankles Elastic 2X-Large
19-167-906 Elastic wrists and ankles Elastic 4X-Large
19-167-907 Elastic wrists and ankles Elastic 5X-Large
19-167-909 Elastic wrists; Attached hood and boots Elastic X-Large
19-167-910 Elastic wrists; Attached hood and boots Elastic 2X-Large
19-167-911 Elastic wrists; Attached hood and boots; Elastic 3X-Large
19-167-030 Knit Collar and Cuffs; Attached hood and boots Knit 4X-Large
19-167-031 Knit Collar and Cuffs; Attached hood and boots Knit 5X-Large
19-167-908 Elastic wrists; Attached hood and boots Elastic Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 19-167-902 Supplier High Five™ Supplier No. EW20103L
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Protection from light splash and particulates
Ideal for general-purpose applications such as maintenance and general cleanup, including oily applications.
  • Microporous construction for breathablity and comfort
  • Full cut for improved fit and comfort
  • Zipper closure
  • Elastic wrists and ankles
  • White
TRUSTED_SUSTAINABILITY
Specifications
Product Type Microporous Coveralls
Size Large
Color White
Cuff Style Elastic
Description Elastic wrists and ankles