Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High Five™ Polyethylene Isolation Gowns

Catalog No. 19156200 Shop All Ansell Products
Change view
Click to view available options
Blue; X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-156-200 Blue; X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-156-200 Supplier High Five™ Supplier No. EB0521400TL
This item is not returnable. View return policy

Impervious to liquid splash and spills

Perfect for protection from liquid splashes and spills

  • Complete with built-in thumb loops and apron-style neck and back yoke
  • Color: Blue

TRUSTED_SUSTAINABILITY

Specifications

Size X-Large
For Use With (Application) Protection from liquid splashes and spills
No. of Pockets None
Gender Unisex
Product Type General Purpose Gown
Material Polyethylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.